missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-chloro-4-nitrophenyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H6340706

 View more versions of this product

Catalog No. AAH6340706



Specifications

Specifications

1-Boc-4-(2-chloro-4-nitrophenyl)piperazine
193902-80-6
10018753
HTMRIXVUKSIEPV-UHFFFAOYSA-N
tert-butyl 4-(2-chloro-4-nitrophenyl)piperazine-1-carboxylate
18783805
Solid
5g
C15H20ClN3O4
tert-butyl 4-2-chloro-4-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-chloro-4-nitrophenyl piperazine, 3-chloro-4-4-t-butoxycarbonyl-1-piperazinyl nitrobenzene, 4-2-chloro-4-nitro-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-chloro-4-nitrophenyl piperazine-1-carboxylic acid t-butyl ester
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
341.792
341.79
97%

Chemical Identifiers

193902-80-6
341.792
tert-butyl 4-2-chloro-4-nitrophenyl piperazine-1-carboxylate, 1-boc-4-2-chloro-4-nitrophenyl piperazine, 3-chloro-4-4-t-butoxycarbonyl-1-piperazinyl nitrobenzene, 4-2-chloro-4-nitro-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-chloro-4-nitrophenyl piperazine-1-carboxylic acid t-butyl ester
tert-butyl 4-(2-chloro-4-nitrophenyl)piperazine-1-carboxylate
C15H20ClN3O4
HTMRIXVUKSIEPV-UHFFFAOYSA-N
18783805
CC(C)(C)OC(=O)N1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
Documents