Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-chloro-4-nitrophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(2-chloro-4-nitrophenyl)piperazine
CAS 193902-80-6
Formula Weight 341.79
Beilstein 10018753
Synonym tert-Butyl 4-(2-chloro-4-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-4-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6340706

 
alfa aesar™
H6340706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6340703

 
alfa aesar™
H6340703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63407MD

 
alfa aesar™
H63407MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 341.79
Beilstein 10018753
Chemical Name or Material 1-Boc-4-(2-chloro-4-nitrophenyl)piperazine
Assay 97%
Physical Form Solid
Quantity 5g
CAS 193902-80-6
Synonym tert-Butyl 4-(2-chloro-4-nitrophenyl)-piperazine-1-carboxylate; 4-(2-Chloro-4-nitrophenyl)-piperazine-1-carboxylic acid tert-butyl ester