Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-4-(2-aminophenyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Boc-4-(2-aminophenyl)piperazine
CAS 170017-74-0
Formula Weight 277.36g/mol
MDL Number MFCD04115046
Synonym tert-Butyl 4-(2-aminophenyl)piperazine-1-carboxylate; 4-(2-Aminophenyl)piperazine-1-carboxylic acid tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6336403

 
alfa aesar™
H63364-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6336406

 
alfa aesar™
H63364-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63364MD

 
alfa aesar™
H63364-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 120 to 122°C
Quantity 1g
Chemical Name or Material 1-Boc-4-(2-aminophenyl)piperazine
CAS 170017-74-0
Physical Form Solid
Assay 97%
Formula Weight 277.36g/mol
MDL Number MFCD04115046
Synonym tert-Butyl 4-(2-aminophenyl)piperazine-1-carboxylate; 4-(2-Aminophenyl)piperazine-1-carboxylic acid tert-butyl ester