Learn More
1-Boc-3-pyrrolidinone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2702303
Description
In the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn the Synthesis and antibacterial evaluation of a novel series of rifabutin-like spirorifamycins 1-Boc-3-pyrrolidinone is used.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
1-Boc-3-pyrrolidinone | |
34°C to 38°C | |
MFCD01631194 | |
n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone | |
JSOMVCDXPUXKIC-UHFFFAOYSA-N | |
tert-butyl 3-oxopyrrolidine-1-carboxylate | |
471360 | |
97% |
101385-93-7 | |
C9H15NO3 | |
1 g | |
Insoluble in water. | |
CC(C)(C)OC(=O)N1CCC(=O)C1 | |
185.223 | |
185.22 |
Chemical Identifiers
101385-93-7 | |
185.223 | |
JSOMVCDXPUXKIC-UHFFFAOYSA-N | |
471360 | |
CC(C)(C)OC(=O)N1CCC(=O)C1 |
C9H15NO3 | |
MFCD01631194 | |
n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone | |
tert-butyl 3-oxopyrrolidine-1-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only