Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-piperidone, 97%

$21.09 - $67.05

Chemical Identifiers

Chemical Name or Material 1-Boc-3-piperidone
CAS 98977-36-7
Molecular Formula C10H17NO3
Formula Weight 199.25
MDL Number MFCD01631193
Beilstein 5936353
Synonym 1-(tert-Butoxycarbonyl)-3-piperidone; tert-Butyl 3-oxo-1-piperidinecarboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1927606

  SDS
alfa aesar™
L1927606
5g Each for $67.05

AAL1927603

  SDS
alfa aesar™
L1927603
1g Each for $21.09

AAL1927614

  SDS
alfa aesar™
L1927614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.25
Chemical Name or Material 1-Boc-3-piperidone
Assay 97%
Molecular Formula C10H17NO3
MDL Number MFCD01631193
CAS 98977-36-7
Melting Point 37° to 39°C
Beilstein 5936353
Synonym 1-(tert-Butoxycarbonyl)-3-piperidone; tert-Butyl 3-oxo-1-piperidinecarboxylate
Quantity 5g

Applications
1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.