Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-iodoindole, 95%

$94.93 - $244.09

Chemical Identifiers

Chemical Name or Material 1-Boc-3-iodoindole
CAS 192189-07-4
Molecular Formula C13H14INO2
Formula Weight 343.16
MDL Number MFCD05864781
Synonym tert-Butyl 3-iodo-1H-indole-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2702203

  SDS
alfa aesar™
H2702203
1g Each for $244.09

AAH27022MD

  SDS
alfa aesar™
H27022MD
250mg Each for $94.93
Description & Specifications

Specifications

Formula Weight 343.16
Chemical Name or Material 1-Boc-3-iodoindole
Assay 95%
Molecular Formula C13H14INO2
MDL Number MFCD05864781
CAS 192189-07-4
Synonym tert-Butyl 3-iodo-1H-indole-1-carboxylate
Quantity 1g
Recommended Storage −30° to −10°C

Applications
1-Boc-3-iodoindole is used as a pharmaceutical intermediate and also used in ceramic manufacturing industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light Sensitive.

Safety and Handling

−30° to −10°C