missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-iodoindole-5-carbonitrile, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 864685-26-7
Molecular Formula C14H13IN2O2
Molecular Weight (g/mol) 368.174
MDL Number MFCD05864767
InChI Key GADLXNFRAAUWAD-UHFFFAOYSA-N
Synonym tert-butyl 5-cyano-3-iodo-1h-indole-1-carboxylate, 1-boc-5-cyano-3-iodo-indole, 1-boc-5-cyano-3-iodoindole, 5-cyano-3-iodoindole-1-carboxylic acid tert-butyl ester, 5-cyano-3-iodo-1h-indole-1-carboxylic acid tert-butyl ester, pubchem7950, 1-tert-butoxycarbonyl-5-cyano-3-iodoindole, tert-butyl 5-cyano-3-iodo-indole-1-carboxylate, tert-butyl 5-cyano-3-iodanyl-indole-1-carboxylate, tert-butyl5-cyano-3-iodo-1h-indole-1-carboxylate
PubChem CID 17749979
IUPAC Name tert-butyl 5-cyano-3-iodoindole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C2=C1C=CC(=C2)C#N)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64169MD View Documents Alfa Aesar™
H64169MD
250mg Each for $28.84
 
AAH6416903 View Documents Alfa Aesar™
H6416903
1g Each for $85.14
 
AAH6416906 View Documents Alfa Aesar™
H6416906
5g Each for $337.73
 
Specifications

Chemical Identifiers

864685-26-7
368.174
GADLXNFRAAUWAD-UHFFFAOYSA-N
17749979
CC(C)(C)OC(=O)N1C=C(C2=C1C=CC(=C2)C#N)I
C14H13IN2O2
MFCD05864767
tert-butyl 5-cyano-3-iodo-1h-indole-1-carboxylate, 1-boc-5-cyano-3-iodo-indole, 1-boc-5-cyano-3-iodoindole, 5-cyano-3-iodoindole-1-carboxylic acid tert-butyl ester, 5-cyano-3-iodo-1h-indole-1-carboxylic acid tert-butyl ester, pubchem7950, 1-tert-butoxycarbonyl-5-cyano-3-iodoindole, tert-butyl 5-cyano-3-iodo-indole-1-carboxylate, tert-butyl 5-cyano-3-iodanyl-indole-1-carboxylate, tert-butyl5-cyano-3-iodo-1h-indole-1-carboxylate
tert-butyl 5-cyano-3-iodoindole-1-carboxylate

Specifications

Light Sensitive
250mg
C14H13IN2O2
UN3439
tert-butyl 5-cyano-3-iodo-1h-indole-1-carboxylate, 1-boc-5-cyano-3-iodo-indole, 1-boc-5-cyano-3-iodoindole, 5-cyano-3-iodoindole-1-carboxylic acid tert-butyl ester, 5-cyano-3-iodo-1h-indole-1-carboxylic acid tert-butyl ester, pubchem7950, 1-tert-butoxycarbonyl-5-cyano-3-iodoindole, tert-butyl 5-cyano-3-iodo-indole-1-carboxylate, tert-butyl 5-cyano-3-iodanyl-indole-1-carboxylate, tert-butyl5-cyano-3-iodo-1h-indole-1-carboxylate
CC(C)(C)OC(=O)N1C=C(C2=C1C=CC(=C2)C#N)I
368.174
368.17
97%
Yellow
864685-26-7
MFCD05864767
20212173
GADLXNFRAAUWAD-UHFFFAOYSA-N
tert-butyl 5-cyano-3-iodoindole-1-carboxylate
17749979
Solid
1-Boc-3-iodoindole-5-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : 2° to 8°C