Please login to your online account to display your discounted pricing

1-BOC-3-iodoazetidine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1-BOC-3-iodoazetidine
Assay Percent Range 95%
Molecular Formula C8H14INO2
Formula Weight 283.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC452080010

  SDS
thermo fisher scientific
AC452080010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC452080050

  SDS
thermo fisher scientific
AC452080050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 283.1
Quantity 1g
Assay Percent Range 95%
Chemical Name or Material 1-BOC-3-iodoazetidine
Molecular Formula C8H14INO2
CAS 254454-54-1