missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-BOC-3-Hydroxyazetidine, 97%, ACROS Organics™

Chemical Identifiers

CAS 141699-55-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
InChI Key XRRXRQJQQKMFBC-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinol, 1-boc-3-hydroxy azetidine, 1-boc-3-hydroxyazetidine, 1-n-boc-3-hydroxyazetidine, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-3-hydroxyazetidine, n-boc-azetidin-3-ol
PubChem CID 2756801
IUPAC Name tert-butyl 3-hydroxyazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441760010
SDS View Documents
Thermo Fisher Scientific
AC441760010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441760050
SDS View Documents
Thermo Fisher Scientific
AC441760050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

141699-55-0
173.21
1-boc-3-azetidinol, 1-boc-3-hydroxy azetidine, 1-boc-3-hydroxyazetidine, 1-n-boc-3-hydroxyazetidine, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-3-hydroxyazetidine, n-boc-azetidin-3-ol
tert-butyl 3-hydroxyazetidine-1-carboxylate
C8H15NO3
XRRXRQJQQKMFBC-UHFFFAOYSA-N
2756801
CC(C)(C)OC(=O)N1CC(C1)O

Specifications

Authentic
White to Yellow
1g
96.0
96% min. (GC)
1-boc-3-azetidinol, 1-boc-3-hydroxy azetidine, 1-boc-3-hydroxyazetidine, 1-n-boc-3-hydroxyazetidine, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-3-hydroxyazetidine, n-boc-azetidin-3-ol
CC(C)(C)OC(=O)N1CC(C1)O
173.21
173.21
97%
Glass Bottle
39.0°C to 44.0°C
141699-55-0
100.0
C8H15NO3
XRRXRQJQQKMFBC-UHFFFAOYSA-N
tert-butyl 3-hydroxyazetidine-1-carboxylate
2756801
Solid
1-BOC-3-Hydroxyazetidine
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS