missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-BOC-3-Hydroxyazetidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441760050

 View more versions of this product

Catalog No. AC441760050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-BOC-3-Hydroxyazetidine
Glass bottle
39.0°C to 44.0°C
141699-55-0
100.0
C8H15NO3
XRRXRQJQQKMFBC-UHFFFAOYSA-N
tert-butyl 3-hydroxyazetidine-1-carboxylate
2756801
Solid
Authentic
White to Yellow
5g
96.0
96% min. (GC)
1-boc-3-azetidinol, 1-boc-3-hydroxy azetidine, 1-boc-3-hydroxyazetidine, 1-n-boc-3-hydroxyazetidine, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-3-hydroxyazetidine, n-boc-azetidin-3-ol
CC(C)(C)OC(=O)N1CC(C1)O
173.21
173.21
97%

Chemical Identifiers

141699-55-0
173.21
1-boc-3-azetidinol, 1-boc-3-hydroxy azetidine, 1-boc-3-hydroxyazetidine, 1-n-boc-3-hydroxyazetidine, 1-tert-butoxycarbonyl-3-hydroxyazetidine, 3-hydroxy-azetidine-1-carboxylic acid tert-butyl ester, 3-hydroxyazetidine-1-carboxylic acid tert-butyl ester, n-boc-3-hydroxy azetidine, n-boc-3-hydroxyazetidine, n-boc-azetidin-3-ol
tert-butyl 3-hydroxyazetidine-1-carboxylate
C8H15NO3
XRRXRQJQQKMFBC-UHFFFAOYSA-N
2756801
CC(C)(C)OC(=O)N1CC(C1)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents