missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-cyanopiperidine, 96%

$132.93 - $468.81

Chemical Identifiers

CAS 91419-53-3
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.277
MDL Number MFCD01861222
InChI Key UEFZTXGFHKPSFS-UHFFFAOYSA-N
Synonym 1-n-boc-3-cyanopiperidine, n-boc-3-cyanopiperidine, 1-boc-3-cyanopiperidine, +/--1-n-boc-3-cyano-piperidine, n-boc-3-cyano-piperidine, 1-n-boc-3-cyano-piperidine, 3-cyano-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-piperidine-3-carbonitrile, 1-tert-butoxycarbonyl-3-cyanopiperidine, tert-butyl-3-cyanopiperidine-1-carboxylate
PubChem CID 2756785
IUPAC Name tert-butyl 3-cyanopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6616403 View Documents Alfa Aesar™
H6616403
1g Each for $132.93
 
AAH6616406 View Documents Alfa Aesar™
H6616406
5g Each for $468.81
 
Specifications

Chemical Identifiers

91419-53-3
210.277
UEFZTXGFHKPSFS-UHFFFAOYSA-N
2756785
CC(C)(C)OC(=O)N1CCCC(C1)C#N
C11H18N2O2
MFCD01861222
1-n-boc-3-cyanopiperidine, n-boc-3-cyanopiperidine, 1-boc-3-cyanopiperidine, +/--1-n-boc-3-cyano-piperidine, n-boc-3-cyano-piperidine, 1-n-boc-3-cyano-piperidine, 3-cyano-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-piperidine-3-carbonitrile, 1-tert-butoxycarbonyl-3-cyanopiperidine, tert-butyl-3-cyanopiperidine-1-carboxylate
tert-butyl 3-cyanopiperidine-1-carboxylate

Specifications

Moisture Sensitive
60°C to 65°C
91419-53-3
MFCD01861222
1-n-boc-3-cyanopiperidine, n-boc-3-cyanopiperidine, 1-boc-3-cyanopiperidine, +/--1-n-boc-3-cyano-piperidine, n-boc-3-cyano-piperidine, 1-n-boc-3-cyano-piperidine, 3-cyano-piperidine-1-carboxylic acid tert-butyl ester, 1-n-boc-piperidine-3-carbonitrile, 1-tert-butoxycarbonyl-3-cyanopiperidine, tert-butyl-3-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)C#N
210.277
210.27
96%
White to Yellow
1g
C11H18N2O2
UN3439
UEFZTXGFHKPSFS-UHFFFAOYSA-N
tert-butyl 3-cyanopiperidine-1-carboxylate
2756785
Powder
1-Boc-3-cyanopiperidine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : 2° to 8°C