Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-cyano-4-pyrrolidinone, 97%

$113.62 - $1,011.04

Chemical Identifiers

Chemical Name or Material 1-Boc-3-cyano-4-pyrrolidinone
CAS 175463-32-8
Molecular Formula C10H14N2O3
Formula Weight 210.23
MDL Number MFCD01318456
Synonym 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidinone; tert-Butyl 3-cyano-4-oxopyrrolidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2930603

  SDS
alfa aesar™
H2930603
1g Each for $307.38

AAH2930606

  SDS
alfa aesar™
H2930606
5g Each for $1,011.04

AAH29306MD

  SDS
alfa aesar™
H29306MD
250mg Each for $113.62
Description & Specifications

Specifications

Formula Weight 210.23
Chemical Name or Material 1-Boc-3-cyano-4-pyrrolidinone
Assay 97%
Molecular Formula C10H14N2O3
Melting Point ≈160°C
MDL Number MFCD01318456
Quantity 1g
CAS 175463-32-8
Synonym 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidinone; tert-Butyl 3-cyano-4-oxopyrrolidine-1-carboxylate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C