Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-azetidinone, 95%

$92.66 - $333.00

Chemical Identifiers

Chemical Name or Material 1-Boc-3-azetidinone
CAS 398489-26-4
Molecular Formula C8H13NO3
Formula Weight 171.2
MDL Number MFCD01861741
Synonym tert-Butyl 3-oxoazetidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5082603

  SDS
alfa aesar™
H5082603
1g Each for $333.00

AAH50826MD

  SDS
alfa aesar™
H50826MD
250mg Each for $92.66
Description & Specifications

Specifications

Formula Weight 171.2
Molecular Formula C8H13NO3
CAS 398489-26-4
Quantity 1g
Synonym tert-Butyl 3-oxoazetidine-1-carboxylate
MDL Number MFCD01861741
Melting Point 49° to 52°C
Assay 95%
Chemical Name or Material 1-Boc-3-azetidinone