Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3-(aminomethyl)pyrrolidine, 95%

$123.55 - $1,137.61

Chemical Identifiers

Chemical Name or Material 1-Boc-3-(aminomethyl)pyrrolidine
CAS 270912-72-6
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD01861798
Synonym 1-tert-Butoxycarbonyl-3-(aminomethyl)pyrrolidine; tert-Butyl 3-(aminomethyl)pyrrolidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2867403

  SDS
alfa aesar™
H2867403
1g Each for $342.04

AAH2867406

  SDS
alfa aesar™
H2867406
5g Each for $1,137.61

AAH28674MD

  SDS
alfa aesar™
H28674MD
250mg Each for $123.55
Description & Specifications

Specifications

Formula Weight 200.28
Assay 95%
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1-Boc-3-(aminomethyl)pyrrolidine
Synonym 1-tert-Butoxycarbonyl-3-(aminomethyl)pyrrolidine; tert-Butyl 3-(aminomethyl)pyrrolidine-1-carboxylate
MDL Number MFCD01861798
Health Hazard 1 Warning
CAS 270912-72-6
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H20N2O2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.