Please login to your online account to display your discounted pricing

1-BOC-3-(aminomethyl)azetidine, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1-BOC-3-(aminomethyl)azetidine
Assay Percent Range 95%
Molecular Formula C9H18N2O2
Formula Weight 186.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC451190010

 
thermo fisher scientific
AC451190010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC451192500

  SDS
thermo fisher scientific
AC451192500
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.25
Quantity 1g
Molecular Formula C9H18N2O2
Chemical Name or Material 1-BOC-3-(aminomethyl)azetidine
CAS 325775-44-8
Assay Percent Range 95%