missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-BOC-3-(aminomethyl)azetidine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451192500
Specifications
1-BOC-3-(aminomethyl)azetidine | |
250mg | |
94% min. (GC) | |
1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester, 1-boc-3-aminomethyl azetidine, 1-boc-3-aminomethyl-azetidine, 1-boc-3-aminomethylazetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl azetidine, n1-boc protected, 3-aminomethyl-1-n-boc-azetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-aminomethyl azetidine-1-carboxylate, tert-butyl 3-aminomethyl azetidinecarboxylate | |
CC(C)(C)OC(=O)N1CC(C1)CN | |
186.25 | |
186.25 |
>110°C | |
325775-44-8 | |
C9H18N2O2 | |
XSJPKMUFBHSIRA-UHFFFAOYSA-N | |
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate | |
10679125 | |
95% |
Chemical Identifiers
325775-44-8 | |
186.25 | |
1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester, 1-boc-3-aminomethyl azetidine, 1-boc-3-aminomethyl-azetidine, 1-boc-3-aminomethylazetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl azetidine, n1-boc protected, 3-aminomethyl-1-n-boc-azetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-aminomethyl azetidine-1-carboxylate, tert-butyl 3-aminomethyl azetidinecarboxylate | |
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate |
C9H18N2O2 | |
XSJPKMUFBHSIRA-UHFFFAOYSA-N | |
10679125 | |
CC(C)(C)OC(=O)N1CC(C1)CN |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air
GHS Signal Word: Warning