Learn More
1-Boc-3-aminoazetidine, 94%, Thermo Scientific Chemicals
$322.23 - $855.67
Chemical Identifiers
CAS | 193269-78-2 |
---|---|
Molecular Formula | C8H16N2O2 |
Molecular Weight (g/mol) | 172.228 |
MDL Number | MFCD01861753 |
InChI Key | WPGLRFGDZJSQGI-UHFFFAOYSA-N |
Synonym | 1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine |
PubChem CID | 1516507 |
IUPAC Name | tert-butyl 3-aminoazetidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CC(C1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH28218MD
|
Thermo Scientific Chemicals
H28218MD |
250 mg |
Each for $322.23
|
|
|||||
AAH2821803
|
Thermo Scientific Chemicals
H2821803 |
1 g |
Each for $855.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
193269-78-2 | |
172.228 | |
WPGLRFGDZJSQGI-UHFFFAOYSA-N | |
1516507 | |
CC(C)(C)OC(=O)N1CC(C1)N |
C8H16N2O2 | |
MFCD01861753 | |
1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine | |
tert-butyl 3-aminoazetidine-1-carboxylate |
Specifications
193269-78-2 | |
MFCD01861753 | |
1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine | |
WPGLRFGDZJSQGI-UHFFFAOYSA-N | |
tert-butyl 3-aminoazetidine-1-carboxylate | |
1516507 | |
94% | |
Air Sensitive |
C8H16N2O2 | |
UN2735 | |
1.465 | |
CC(C)(C)OC(=O)N1CC(C1)N | |
172.228 | |
172.23 | |
250 mg | |
1-Boc-3-aminoazetidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only