Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-3,3-difluoropyrrolidine, 98%

$87.08 - $223.00

Chemical Identifiers

Chemical Name or Material 1-Boc-3,3-difluoropyrrolidine
CAS 195447-25-7
Molecular Formula C9H15F2NO2
Formula Weight 207.22
MDL Number MFCD06797010
Synonym tert-Butyl 3,3-difluoropyrroline-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2742203

  SDS
alfa aesar™
H2742203
1g Each for $223.00

AAH27422MD

  SDS
alfa aesar™
H27422MD
250mg Each for $87.08
Description & Specifications

Specifications

Formula Weight 207.22
Assay 98%
Quantity 1g
Molecular Formula C9H15F2NO2
Synonym tert-Butyl 3,3-difluoropyrroline-1-carboxylate
MDL Number MFCD06797010
Chemical Name or Material 1-Boc-3,3-difluoropyrrolidine
CAS 195447-25-7