missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-2-piperidone, 99%

$41.18 - $110.57

Chemical Identifiers

CAS 85908-96-9
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD02179046
InChI Key ULMHMJAEGZPQRY-UHFFFAOYSA-N
Synonym 1-boc-2-piperidone, n-boc-2-piperidone, 1-boc-1-azacyclohexan-2-one, 1-tert-butoxycarbonyl-2-piperidone, n-tert-butoxycarbonyl-2-piperidone, tert-butyl 2-oxo-1-piperidinecarboxylate, 1-piperidinecarboxylic acid, 2-oxo-, 1,1-dimethylethyl ester, tert-butyl 2-oxopiperidinecarboxylate, boc-piperidinone, n-boc piperidone
PubChem CID 7577838
ChEBI CHEBI:59556
IUPAC Name tert-butyl 2-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1889603 View Documents Alfa Aesar™
L1889603
1g Each for $41.18
 
AAL1889606 View Documents Alfa Aesar™
L1889606
5g Each for $110.57
 
Specifications

Chemical Identifiers

85908-96-9
199.25
ULMHMJAEGZPQRY-UHFFFAOYSA-N
7577838
tert-butyl 2-oxopiperidine-1-carboxylate
C10H17NO3
MFCD02179046
1-boc-2-piperidone, n-boc-2-piperidone, 1-boc-1-azacyclohexan-2-one, 1-tert-butoxycarbonyl-2-piperidone, n-tert-butoxycarbonyl-2-piperidone, tert-butyl 2-oxo-1-piperidinecarboxylate, 1-piperidinecarboxylic acid, 2-oxo-, 1,1-dimethylethyl ester, tert-butyl 2-oxopiperidinecarboxylate, boc-piperidinone, n-boc piperidone
CHEBI:59556
CC(C)(C)OC(=O)N1CCCCC1=O

Specifications

34°C to 37°C
85908-96-9
MFCD02179046
1-boc-2-piperidone, n-boc-2-piperidone, 1-boc-1-azacyclohexan-2-one, 1-tert-butoxycarbonyl-2-piperidone, n-tert-butoxycarbonyl-2-piperidone, tert-butyl 2-oxo-1-piperidinecarboxylate, 1-piperidinecarboxylic acid, 2-oxo-, 1,1-dimethylethyl ester, tert-butyl 2-oxopiperidinecarboxylate, boc-piperidinone, n-boc piperidone
CC(C)(C)OC(=O)N1CCCCC1=O
199.25
CHEBI:59556
99%
1g
C10H17NO3
4674955
ULMHMJAEGZPQRY-UHFFFAOYSA-N
tert-butyl 2-oxopiperidine-1-carboxylate
7577838
199.25
1-Boc-2-piperidone