Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Boc-2-(hydroxydimethylsilyl)pyrrole, 97%

$366.35 - $1,108.28

Chemical Identifiers

Chemical Name or Material 1-Boc-2-(hydroxydimethylsilyl)pyrrole
CAS 879904-82-2
Molecular Formula C11H19NO3Si
Formula Weight 241.36
MDL Number MFCD08457657
Synonym tert-Butyl 2-(hydroxydimethylsilyl)pyrrole-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2842803

 
alfa aesar™
H2842803
1g Each for $366.35

AAH2842806

 
alfa aesar™
H2842806
5g Each for $1,108.28
Description & Specifications

Specifications

Formula Weight 241.36
Chemical Name or Material 1-Boc-2-(hydroxydimethylsilyl)pyrrole
Assay 97%
Molecular Formula C11H19NO3Si
Melting Point 53° to 57°C
MDL Number MFCD08457657
Quantity 1g
CAS 879904-82-2
Synonym tert-Butyl 2-(hydroxydimethylsilyl)pyrrole-1-carboxylate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C