Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-Boc-2-(aminomethyl)pyrrolidine, tech. 85%

Chemical Identifiers

Chemical Name or Material (+/-)-1-Boc-2-(aminomethyl)pyrrolidine
CAS 177911-87-4
Purity Grade Technical Grade
Molecular Formula C10H20N2O2
Formula Weight 200.28
MDL Number MFCD01317793
UN Number UN2735
Synonym (+/-)-1-tert-Butoxycarbonyl-2-(aminomethyl)pyrrolidine; tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2865006

 
alfa aesar™
H2865006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2865003

 
alfa aesar™
H2865003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH28650MD

 
alfa aesar™
H28650MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.28
Chemical Name or Material (+/-)-1-Boc-2-(aminomethyl)pyrrolidine
Purity Grade Technical Grade
Assay 85%
Molecular Formula C10H20N2O2
MDL Number MFCD01317793
Quantity 5g
CAS 177911-87-4
UN Number UN2735
Synonym (+/-)-1-tert-Butoxycarbonyl-2-(aminomethyl)pyrrolidine; tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.