missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Boc-2,6-diazaspiro[3.3]heptane hemioxalate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6376303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
1-Boc-2,6-diazaspiro[3.3]heptane hemioxalate | |
C22H38N4O8 | |
tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate oxalate 2:1, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxylate, tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate hemioxalate, tert-butyl 2,6-diazaspiro 3,3 heptane-2-carboxylate hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid, 1,1-dimethylethyl ester, ethanedioate 2:1, oxalic acid; bis tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate, 2-boc-2,6-diazaspiro 3.3 heptaneoxalate, 2-boc-2,6-diazaspiro 3.3 heptane hemioxalate, 1-boc-2,6-diazaspiro 3.3 heptane hemioxalate | |
PYRAMVPOMDHTNX-UHFFFAOYSA-N | |
tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate;oxalic acid | |
486.566 | |
441.54 | |
Solid |
1041026-71-4 | |
MFCD12404932 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)N1CC2(C1)CNC2.CC(C)(C)OC(=O)N1CC2(C1)CNC2.C(=O)(C(=O)O)O | |
1 g | |
50902214 | |
97% |
Chemical Identifiers
1041026-71-4 | |
486.566 | |
PYRAMVPOMDHTNX-UHFFFAOYSA-N | |
50902214 | |
CC(C)(C)OC(=O)N1CC2(C1)CNC2.CC(C)(C)OC(=O)N1CC2(C1)CNC2.C(=O)(C(=O)O)O |
C22H38N4O8 | |
MFCD12404932 | |
tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate oxalate 2:1, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxylate, tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate hemioxalate, tert-butyl 2,6-diazaspiro 3,3 heptane-2-carboxylate hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid, 1,1-dimethylethyl ester, ethanedioate 2:1, oxalic acid; bis tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate, 2-boc-2,6-diazaspiro 3.3 heptaneoxalate, 2-boc-2,6-diazaspiro 3.3 heptane hemioxalate, 1-boc-2,6-diazaspiro 3.3 heptane hemioxalate | |
tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate;oxalic acid |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
TSCA : No
Recommended Storage : Ambient temperatures