missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Boc-2,6-diazaspiro[3.3]heptane hemioxalate, 97%, Alfa Aesar™

$239.03 - $760.27

Chemical Identifiers

CAS 1041026-71-4
Molecular Formula C22H38N4O8
Molecular Weight (g/mol) 486.566
MDL Number MFCD12404932
InChI Key PYRAMVPOMDHTNX-UHFFFAOYSA-N
Synonym tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate oxalate 2:1, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxylate, tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate hemioxalate, tert-butyl 2,6-diazaspiro 3,3 heptane-2-carboxylate hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid, 1,1-dimethylethyl ester, ethanedioate 2:1, oxalic acid; bis tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate, 2-boc-2,6-diazaspiro 3.3 heptaneoxalate, 2-boc-2,6-diazaspiro 3.3 heptane hemioxalate, 1-boc-2,6-diazaspiro 3.3 heptane hemioxalate
PubChem CID 50902214
IUPAC Name tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate;oxalic acid
SMILES CC(C)(C)OC(=O)N1CC2(C1)CNC2.CC(C)(C)OC(=O)N1CC2(C1)CNC2.C(=O)(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63763MD View Documents Alfa Aesar
H63763MD
250mg Each for $239.03
 
AAH6376303 View Documents Alfa Aesar
H6376303
1g Each for $760.27
 
Description

Description

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1041026-71-4
486.566
PYRAMVPOMDHTNX-UHFFFAOYSA-N
50902214
CC(C)(C)OC(=O)N1CC2(C1)CNC2.CC(C)(C)OC(=O)N1CC2(C1)CNC2.C(=O)(C(=O)O)O
C22H38N4O8
MFCD12404932
tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate oxalate 2:1, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxylate, tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate hemioxalate, tert-butyl 2,6-diazaspiro 3,3 heptane-2-carboxylate hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid, 1,1-dimethylethyl ester, ethanedioate 2:1, oxalic acid; bis tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate, 2-boc-2,6-diazaspiro 3.3 heptaneoxalate, 2-boc-2,6-diazaspiro 3.3 heptane hemioxalate, 1-boc-2,6-diazaspiro 3.3 heptane hemioxalate
tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate;oxalic acid

Specifications

250mg
C22H38N4O8
tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate oxalate 2:1, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxylate, tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate hemioxalate, tert-butyl 2,6-diazaspiro 3,3 heptane-2-carboxylate hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester hemioxalate, 2,6-diazaspiro 3.3 heptane-2-carboxylic acid, 1,1-dimethylethyl ester, ethanedioate 2:1, oxalic acid; bis tert-butyl 2,6-diazaspiro 3.3 heptane-2-carboxylate, 2-boc-2,6-diazaspiro 3.3 heptaneoxalate, 2-boc-2,6-diazaspiro 3.3 heptane hemioxalate, 1-boc-2,6-diazaspiro 3.3 heptane hemioxalate
CC(C)(C)OC(=O)N1CC2(C1)CNC2.CC(C)(C)OC(=O)N1CC2(C1)CNC2.C(=O)(C(=O)O)O
486.566
441.54
97%
1041026-71-4
MFCD12404932
PYRAMVPOMDHTNX-UHFFFAOYSA-N
tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate;oxalic acid
50902214
Solid
1-Boc-2,6-diazaspiro[3.3]heptane hemioxalate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning