missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[Bis(4-fluorophenyl)methyl]piperazine, TCI America™

$99.31 - $339.17

Chemical Identifiers

CAS 27469-60-9
Molecular Formula C17H18F2N2
Molecular Weight (g/mol) 288.342
InChI Key TTXIFFYPVGWLSE-UHFFFAOYSA-N
Synonym 1-bis 4-fluorophenyl methyl piperazine, 1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 1-bis 4-fluorophenyl methyl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
PubChem CID 152932
IUPAC Name 1-[bis(4-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27645G View Documents TCI America
B27645G
5g Each for $99.31
 
B276425G View Documents TCI America
B276425G
25g Each for $339.17
 
Specifications

Chemical Identifiers

27469-60-9
288.342
1-bis 4-fluorophenyl methyl piperazine, 1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 1-bis 4-fluorophenyl methyl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
1-[bis(4-fluorophenyl)methyl]piperazine
C17H18F2N2
TTXIFFYPVGWLSE-UHFFFAOYSA-N
152932
C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F

Specifications

130°C
93°C
27469-60-9
2811
TTXIFFYPVGWLSE-UHFFFAOYSA-N
1-[bis(4-fluorophenyl)methyl]piperazine
152932
Crystal-Powder at 20°C
1-[Bis(4-fluorophenyl)methyl]piperazine
White-Yellow
5g
C17H18F2N2
1-bis 4-fluorophenyl methyl piperazine, 1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 1-bis 4-fluorophenyl methyl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
288.342
288.34
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No