missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[Bis(4-fluorophenyl)methyl]piperazine, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5517306

 View more versions of this product

Catalog No. AAH5517306



Specifications

Specifications

1-[Bis(4-fluorophenyl)methyl]piperazine
5g
C17H18F2N2
UN2811
1 bis 4-fluorophenyl methyl-piperazine
C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
288.342
288.34
88°C to 92°C
27469-60-9
MFCD00038660
620754
TTXIFFYPVGWLSE-UHFFFAOYSA-N
1-[bis(4-fluorophenyl)methyl]piperazine
152932
97%

Chemical Identifiers

27469-60-9
288.342
TTXIFFYPVGWLSE-UHFFFAOYSA-N
152932
C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
C17H18F2N2
MFCD00038660
1 bis 4-fluorophenyl methyl-piperazine
1-[bis(4-fluorophenyl)methyl]piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 248-476-0

TSCA : No

Recommended Storage : Ambient temperatures

Documents