missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[Bis(2-hydroxyethyl)amino]-2-propanol, TCI America™

$79.45 - $275.00

Chemical Identifiers

CAS 6712-98-7
Molecular Formula C7H17NO3
Molecular Weight (g/mol) 163.217
MDL Number MFCD00009725
InChI Key ZFECCYLNALETDE-UHFFFAOYSA-N
Synonym 1-bis 2-hydroxyethyl amino-2-propanol, 1-bis 2-hydroxyethyl amino propan-2-ol, 2-propanol, 1-bis 2-hydroxyethyl amino, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, n,n-bis 2-hydroxyethyl isopropanolamine, 2,2'-2-hydroxypropyl azanediyl diethanol, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, 2-propanol, 1-bis 2-hydroxyethyl amino, acmc-209nxu, 1-n,n-bis 2-hydroxyethyl amino propan-2-ol
PubChem CID 97712
IUPAC Name 1-[bis(2-hydroxyethyl)amino]propan-2-ol
SMILES CC(CN(CCO)CCO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21805G View Documents TCI America
B21805G
5g Each for $79.45
 
B218025G View Documents TCI America
B218025G
25g Each for $275.00
 
Specifications

Chemical Identifiers

6712-98-7
163.217
ZFECCYLNALETDE-UHFFFAOYSA-N
97712
CC(CN(CCO)CCO)O
C7H17NO3
MFCD00009725
1-bis 2-hydroxyethyl amino-2-propanol, 1-bis 2-hydroxyethyl amino propan-2-ol, 2-propanol, 1-bis 2-hydroxyethyl amino, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, n,n-bis 2-hydroxyethyl isopropanolamine, 2,2'-2-hydroxypropyl azanediyl diethanol, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, 2-propanol, 1-bis 2-hydroxyethyl amino, acmc-209nxu, 1-n,n-bis 2-hydroxyethyl amino propan-2-ol
1-[bis(2-hydroxyethyl)amino]propan-2-ol

Specifications

145°C
5g
C7H17NO3
1-bis 2-hydroxyethyl amino-2-propanol, 1-bis 2-hydroxyethyl amino propan-2-ol, 2-propanol, 1-bis 2-hydroxyethyl amino, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, n,n-bis 2-hydroxyethyl isopropanolamine, 2,2'-2-hydroxypropyl azanediyl diethanol, 1-n,n-bis 2-hydroxyethyl amino-2-propanol, 2-propanol, 1-bis 2-hydroxyethyl amino, acmc-209nxu, 1-n,n-bis 2-hydroxyethyl amino propan-2-ol
CC(CN(CCO)CCO)O
163.217
163.22
≥93.0% (GC)
Yellow
6712-98-7
MFCD00009725
ZFECCYLNALETDE-UHFFFAOYSA-N
1-[bis(2-hydroxyethyl)amino]propan-2-ol
97712
Clear Liquid-Slightly Cloudy Liquid at 20°C
1-[Bis(2-hydroxyethyl)amino]-2-propanol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0353

TSCA : Yes