missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzylpyrrole, 97%

Manufacturer:  Alfa Aesar™ H5593606

 View more versions of this product

Catalog No. AAH5593606



Specifications

Specifications

1-Benzylpyrrole
1.5685
Colorless to Brown
2051-97-0
MFCD00963301
1-benzyl-1h-pyrrole
C1=CC=C(C=C1)CN2C=CC=C2
157.216
157.21
97%
1.020 g/mL
90°C (0.9mmHg)
5g
C11H11N
UN1993
FNEQHKCQXDKYEO-UHFFFAOYSA-N
1-benzylpyrrole
74922
Liquid

Chemical Identifiers

2051-97-0
157.216
FNEQHKCQXDKYEO-UHFFFAOYSA-N
74922
C1=CC=C(C=C1)CN2C=CC=C2
C11H11N
MFCD00963301
1-benzyl-1h-pyrrole
1-benzylpyrrole
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 218-137-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents