Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzylpiperidine-4-carboxaldehyde, 95%

$134.23 - $438.69

Chemical Identifiers

Chemical Name or Material 1-Benzylpiperidine-4-carboxaldehyde
CAS 22065-85-6
Molecular Formula C13H17NO
Formula Weight 203.28
MDL Number MFCD02258902
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2702103

 
alfa aesar™
H2702103
1g Each for $134.23

AAH2702106

 
alfa aesar™
H2702106
5g Each for $438.69
Description & Specifications

Specifications

Formula Weight 203.28
Chemical Name or Material 1-Benzylpiperidine-4-carboxaldehyde
Assay 95%
Molecular Formula C13H17NO
MDL Number MFCD02258902
CAS 22065-85-6
Boiling Point 315°C
Health Hazard 1 Warning
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C