Please login to your online account to display your discounted pricing

1-Benzylpiperidin-4-amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzylpiperidin-4-amine
CAS 50541-93-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H18N2
Formula Weight 190.29
MDL Number MFCD00006504
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01177DA

  SDS
fisher scientific company
SB01177DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Formula Weight 190.29
Assay 97%
Density 0.9300g/mL
CAS Min % 97.0
Packaging Amber Glass Bottle
CAS 50541-93-0
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C12H18N2
MDL Number MFCD00006504
Chemical Name or Material 1-Benzylpiperidin-4-amine
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.