Please login to your online account to display your discounted pricing

1-Benzylpiperidin-4-amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SB01177DA

 View more versions of this product

Catalog No. SB01177DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1g
Assay 97%
Formula Weight 190.29
Density 0.9300g/mL
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H18N2
MDL Number MFCD00006504
CAS 50541-93-0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Benzylpiperidin-4-amine
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 1-Benzylpiperidin-4-amine
CAS 50541-93-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H18N2
Formula Weight 190.29
MDL Number MFCD00006504
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

SDS & Safety Documents