missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonylmethyl-N-Boc-D-histidine, 98%

$18.04 - $232.86

Chemical Identifiers

CAS 99310-01-7
Molecular Formula C19H25N3O5
Molecular Weight (g/mol) 375.425
MDL Number MFCD00076940
InChI Key LPVKZCHCZSFTOJ-MRXNPFEDSA-N
Synonym boc-d-his bom-oh, 2r-3-3-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-his 3-bom-oh, boc-d-his-bom-oh, n, a-boc-, eth-bom-d-histidine, boc-np-benzyloxymethyl-d-histidine, n, a-boc-n im-benzyloxymethyl-d-histidine, boc-d-his bom-oh tlc
PubChem CID 7018759
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[3-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CN1COCC2=CC=CC=C2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6633903 View Documents Alfa Aesar™
H6633903
1g Each for $18.04
 
AAH6633906 View Documents Alfa Aesar™
H6633906
5g Each for $59.05
 
AAH6633914 View Documents Alfa Aesar™
H6633914
25g Each for $232.86
 
Specifications

Chemical Identifiers

99310-01-7
375.425
LPVKZCHCZSFTOJ-MRXNPFEDSA-N
7018759
CC(C)(C)OC(=O)NC(CC1=CN=CN1COCC2=CC=CC=C2)C(=O)O
C19H25N3O5
MFCD00076940
boc-d-his bom-oh, 2r-3-3-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-his 3-bom-oh, boc-d-his-bom-oh, n, a-boc-, eth-bom-d-histidine, boc-np-benzyloxymethyl-d-histidine, n, a-boc-n im-benzyloxymethyl-d-histidine, boc-d-his bom-oh tlc
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[3-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid

Specifications

99310-01-7
MFCD00076940
LPVKZCHCZSFTOJ-MRXNPFEDSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[3-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
7018759
Powder
White
1-Benzyloxycarbonylmethyl-N-Boc-D-histidine
C19H25N3O5
boc-d-his bom-oh, 2r-3-3-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-his 3-bom-oh, boc-d-his-bom-oh, n, a-boc-, eth-bom-d-histidine, boc-np-benzyloxymethyl-d-histidine, n, a-boc-n im-benzyloxymethyl-d-histidine, boc-d-his bom-oh tlc
CC(C)(C)OC(=O)NC(CC1=CN=CN1COCC2=CC=CC=C2)C(=O)O
375.425
375.43
98%
1g