missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid, 97%

$53.05 - $766.43

Chemical Identifiers

CAS 190004-53-6
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD03426163
InChI Key GDJSFBNRXFOUEQ-UHFFFAOYSA-N
Synonym 1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, 1-z-amino cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid, z-ac4c-oh, 1-benzyloxycarbonylamino-cyclobutyl-1-carboxylic acid, 1-benzyloxy carbonyl amino-cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, z-cyclovaline, z-acbc-oh, 1-benzyloxycarbonylaminocyclobutanecarboxylic acid
PubChem CID 1512648
IUPAC Name 1-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
SMILES C1CC(C1)(C(=O)O)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62122MD View Documents Alfa Aesar™
H62122MD
250mg Each for $53.05
 
AAH6212203 View Documents Alfa Aesar™
H6212203
1g Each for $168.09
 
AAH6212206 View Documents Alfa Aesar™
H6212206
5g Each for $766.43
 
Specifications

Chemical Identifiers

190004-53-6
249.266
GDJSFBNRXFOUEQ-UHFFFAOYSA-N
1512648
C1CC(C1)(C(=O)O)NC(=O)OCC2=CC=CC=C2
C13H15NO4
MFCD03426163
1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, 1-z-amino cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid, z-ac4c-oh, 1-benzyloxycarbonylamino-cyclobutyl-1-carboxylic acid, 1-benzyloxy carbonyl amino-cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, z-cyclovaline, z-acbc-oh, 1-benzyloxycarbonylaminocyclobutanecarboxylic acid
1-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid

Specifications

83°C to 87°C
190004-53-6
MFCD03426163
GDJSFBNRXFOUEQ-UHFFFAOYSA-N
1-(phenylmethoxycarbonylamino)cyclobutane-1-carboxylic acid
1512648
97%
250mg
C13H15NO4
1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, 1-z-amino cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutane-1-carboxylic acid, z-ac4c-oh, 1-benzyloxycarbonylamino-cyclobutyl-1-carboxylic acid, 1-benzyloxy carbonyl amino-cyclobutanecarboxylic acid, 1-benzyloxy carbonyl amino cyclobutanecarboxylic acid, z-cyclovaline, z-acbc-oh, 1-benzyloxycarbonylaminocyclobutanecarboxylic acid
C1CC(C1)(C(=O)O)NC(=O)OCC2=CC=CC=C2
249.266
249.26
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning