missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid, 97%

$20.36 - $260.62

Chemical Identifiers

CAS 188527-21-1
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD02179036
InChI Key JSASVUTVTRNJHA-UHFFFAOYSA-N
Synonym 1-n-cbz-pyrrolidine-3-carboxylic acid, n-cbz-pyrrolidine-3-carboxylic acid, 1-benzyloxycarbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-cbz-pyrrolidine-3-carboxylic acid, 1-n-cbz-beta-proline, 1-n-cbz-belta-proline, 1-cbz-3-pyrrolidinecarboxylic acid, 3-carboxy-1-cbz-pyrrolidine
PubChem CID 18538483
IUPAC Name 1-phenylmethoxycarbonylpyrrolidine-3-carboxylic acid
SMILES C1CN(CC1C(=O)O)C(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64728MD View Documents Alfa Aesar™
H64728MD
250mg Each for $20.36
 
AAH6472803 View Documents Alfa Aesar™
H6472803
1g Each for $65.54
 
AAH6472806 View Documents Alfa Aesar™
H6472806
5g Each for $260.62
 
Specifications

Chemical Identifiers

188527-21-1
249.266
JSASVUTVTRNJHA-UHFFFAOYSA-N
18538483
C1CN(CC1C(=O)O)C(=O)OCC2=CC=CC=C2
C13H15NO4
MFCD02179036
1-n-cbz-pyrrolidine-3-carboxylic acid, n-cbz-pyrrolidine-3-carboxylic acid, 1-benzyloxycarbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-cbz-pyrrolidine-3-carboxylic acid, 1-n-cbz-beta-proline, 1-n-cbz-belta-proline, 1-cbz-3-pyrrolidinecarboxylic acid, 3-carboxy-1-cbz-pyrrolidine
1-phenylmethoxycarbonylpyrrolidine-3-carboxylic acid

Specifications

250mg
C13H15NO4
13268300
JSASVUTVTRNJHA-UHFFFAOYSA-N
1-phenylmethoxycarbonylpyrrolidine-3-carboxylic acid
18538483
Solid
1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid
188527-21-1
MFCD02179036
1-n-cbz-pyrrolidine-3-carboxylic acid, n-cbz-pyrrolidine-3-carboxylic acid, 1-benzyloxycarbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-cbz-pyrrolidine-3-carboxylic acid, 1-n-cbz-beta-proline, 1-n-cbz-belta-proline, 1-cbz-3-pyrrolidinecarboxylic acid, 3-carboxy-1-cbz-pyrrolidine
C1CN(CC1C(=O)O)C(=O)OCC2=CC=CC=C2
249.266
249.27
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning