Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonyl-L-prolinamide, 95%

Chemical Identifiers

Chemical Name or Material 1-Benzyloxycarbonyl-L-prolinamide
CAS 34079-31-7
Formula Weight 248.28
MDL Number MFCD00020830
Beilstein 88577
Synonym Z-Pro-NH2; Z-L-Prolinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6530706

  SDS
alfa aesar™
H6530706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6530714

  SDS
alfa aesar™
H6530714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6530722

  SDS
alfa aesar™
H6530722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 90° to 95°C
Formula Weight 248.28
Chemical Name or Material 1-Benzyloxycarbonyl-L-prolinamide
Assay 95%
MDL Number MFCD00020830
CAS 34079-31-7
Beilstein 88577
Physical Form Solid
Storage Note 1 2° to 8°C
Quantity 5g
Synonym Z-Pro-NH2; Z-L-Prolinamide
Safety and Handling

2° to 8°C