Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonyl-4-piperidone, 98+%

$20.32 - $20.32

Chemical Identifiers

Chemical Name or Material 1-Benzyloxycarbonyl-4-piperidone
CAS 19099-93-5
Molecular Formula C13H15NO3
Formula Weight 233.27
MDL Number MFCD00673144
Synonym Benzyl 4-oxo-1-piperidinecarboxylate; N-Cbz-4-piperidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2318103

  SDS
alfa aesar™
B2318103
1g Each for $20.32
Description & Specifications

Specifications

Formula Weight 233.27
Chemical Name or Material 1-Benzyloxycarbonyl-4-piperidone
Assay ≥98%
Molecular Formula C13H15NO3
MDL Number MFCD00673144
CAS 19099-93-5
Synonym Benzyl 4-oxo-1-piperidinecarboxylate; N-Cbz-4-piperidone
Melting Point 38° to 41°C
Health Hazard 1 Warning
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.