missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonyl-4-oxo-L-proline, 97%

$54.12 - $737.14

Chemical Identifiers

CAS 64187-47-9
Molecular Formula C13H13NO5
Molecular Weight (g/mol) 263.249
MDL Number MFCD07367987
InChI Key RPLLCMZOIFOBIF-NSHDSACASA-N
Synonym n-carbobenzyloxy-4-keto-l-proline, n-carbobenzoxy-4-oxo-l-proline, s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-cbz-4-oxo-l-proline, s-1-z-4-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 2s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, 2s-4-oxo-1,2-pyrrolidinedicarboxylic acid 1-phenylmethyl ester, n-cbz-4-ketoproline, pubchem14286
PubChem CID 11076282
IUPAC Name (2S)-4-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
SMILES C1C(N(CC1=O)C(=O)OCC2=CC=CC=C2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66054MD View Documents Alfa Aesar™
H66054MD
250mg Each for $54.12
 
AAH6605403 View Documents Alfa Aesar™
H6605403
1g Each for $165.01
 
AAH6605406 View Documents Alfa Aesar™
H6605406
5g Each for $737.14
 
Specifications

Chemical Identifiers

64187-47-9
263.249
RPLLCMZOIFOBIF-NSHDSACASA-N
11076282
C1C(N(CC1=O)C(=O)OCC2=CC=CC=C2)C(=O)O
C13H13NO5
MFCD07367987
n-carbobenzyloxy-4-keto-l-proline, n-carbobenzoxy-4-oxo-l-proline, s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-cbz-4-oxo-l-proline, s-1-z-4-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 2s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, 2s-4-oxo-1,2-pyrrolidinedicarboxylic acid 1-phenylmethyl ester, n-cbz-4-ketoproline, pubchem14286
(2S)-4-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid

Specifications

64187-47-9
MFCD07367987
RPLLCMZOIFOBIF-NSHDSACASA-N
(2S)-4-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
11076282
Powder
Brown to White
250mg
C13H13NO5
n-carbobenzyloxy-4-keto-l-proline, n-carbobenzoxy-4-oxo-l-proline, s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-cbz-4-oxo-l-proline, s-1-z-4-oxopyrrolidine-2-carboxylic acid, s-1-benzyloxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 2s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, 2s-4-oxo-1,2-pyrrolidinedicarboxylic acid 1-phenylmethyl ester, n-cbz-4-ketoproline, pubchem14286
C1C(N(CC1=O)C(=O)OCC2=CC=CC=C2)C(=O)O
263.249
263.24
97%
95°C to 100°C
1-Benzyloxycarbonyl-4-oxo-L-proline