Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid, 97%

$96.51 - $731.16

Chemical Identifiers

Chemical Name or Material (+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid
CAS 126937-41-5
Molecular Formula C18H24N2O6
Formula Weight 364.4
MDL Number MFCD01631192
Synonym (+/-)-4-Boc-1-Z-piperazine-2-carboxylic acid; (+/-)-Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl diester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2704703

  SDS
alfa aesar™
H2704703
1g Each for $241.66

AAH2704706

  SDS
alfa aesar™
H2704706
5g Each for $731.16

AAH27047MD

  SDS
alfa aesar™
H27047MD
250mg Each for $96.51
Description & Specifications

Specifications

Formula Weight 364.4
Chemical Name or Material (+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid
Assay 97%
Molecular Formula C18H24N2O6
MDL Number MFCD01631192
CAS 126937-41-5
Synonym (+/-)-4-Boc-1-Z-piperazine-2-carboxylic acid; (+/-)-Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl diester
Melting Point 150° to 152°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.