Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonyl-3-piperidone, ≥97%

Chemical Identifiers

Chemical Name or Material 1-Benzyloxycarbonyl-3-piperidone
CAS 61995-20-8
Formula Weight 233.27
MDL Number MFCD03001723
Beilstein 1464624
Synonym Benzyl 3-oxopiperidine-1-carboxylate; 1-N-Cbz-3-piperidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6447103

 
alfa aesar™
H6447103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6447106

 
alfa aesar™
H6447106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6447114

 
alfa aesar™
H6447114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.27
Chemical Name or Material 1-Benzyloxycarbonyl-3-piperidone
Physical Form Liquid
Assay ≥97%
MDL Number MFCD03001723
CAS 61995-20-8
Beilstein 1464624
Quantity 1g
Synonym Benzyl 3-oxopiperidine-1-carboxylate; 1-N-Cbz-3-piperidone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C

Applications
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C