Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonyl-3-bromopyrrolidine, tech. 90%

Chemical Identifiers

Chemical Name or Material 1-Benzyloxycarbonyl-3-bromopyrrolidine
CAS 220212-12-4
Purity Grade Technical Grade
Formula Weight 284.15
MDL Number MFCD13181593
Synonym 1-Cbz-3-bromopyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6667603

  SDS
alfa aesar™
H6667603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6667606

  SDS
alfa aesar™
H6667606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66676MD

  SDS
alfa aesar™
H66676MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 284.15
Chemical Name or Material 1-Benzyloxycarbonyl-3-bromopyrrolidine
Assay 90%
MDL Number MFCD13181593
Synonym 1-Cbz-3-bromopyrrolidine
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.5570
Purity Grade Technical Grade
Physical Form Liquid
Density 1.395g/mL
CAS 220212-12-4
Color Yellow to dark brown
Health Hazard 1 Warning
Quantity 1g
Storage Note 1 2° to 8°C

Applications
A commonly used as an organic reagent and intermediate for pharmaceutical.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C