Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine, 97%

Chemical Identifiers

Chemical Name or Material 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine
Molecular Formula C13H13FN2O2
Formula Weight 248.25
MDL Number MFCD08458685
UN Number UN3439
Synonym Benzyl (2S,4R)-2-cyano-4-fluoro-pyrrolidine-1-carboxylate; N-Cbz-trans-4-fluoro-L-prolinonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2758303

  SDS
alfa aesar™
H2758303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH27583MD

  SDS
alfa aesar™
H27583MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.25
Assay 97%
Quantity 1g
Melting Point 68° to 72°C
Molecular Formula C13H13FN2O2
Synonym Benzyl (2S,4R)-2-cyano-4-fluoro-pyrrolidine-1-carboxylate; N-Cbz-trans-4-fluoro-L-prolinonitrile
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 1 Danger
MDL Number MFCD08458685
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3439
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Chemical Name or Material 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.