missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxy-2-methyl-3-nitrobenzene, 98%

$36.64 - $519.12

Chemical Identifiers

CAS 20876-37-3
Molecular Formula C14H13NO3
Molecular Weight (g/mol) 243.262
MDL Number MFCD00100652
InChI Key PBSZHNXXFIYDBU-UHFFFAOYSA-N
Synonym 1-benzyloxy-2-methyl-3-nitrobenzene
PubChem CID 292648
IUPAC Name 2-methyl-1-nitro-3-phenylmethoxybenzene
SMILES CC1=C(C=CC=C1OCC2=CC=CC=C2)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5589903 View Documents Alfa Aesar™
H5589903
1g Each for $36.64
 
AAH5589906 View Documents Alfa Aesar™
H5589906
5g Each for $138.70
 
AAH5589914 View Documents Alfa Aesar™
H5589914
25g Each for $519.12
 
Specifications

Chemical Identifiers

20876-37-3
243.262
PBSZHNXXFIYDBU-UHFFFAOYSA-N
292648
CC1=C(C=CC=C1OCC2=CC=CC=C2)[N+](=O)[O-]
C14H13NO3
MFCD00100652
1-benzyloxy-2-methyl-3-nitrobenzene
2-methyl-1-nitro-3-phenylmethoxybenzene

Specifications

Brown to Yellow
1g
C14H13NO3
1-benzyloxy-2-methyl-3-nitrobenzene
CC1=C(C=CC=C1OCC2=CC=CC=C2)[N+](=O)[O-]
243.262
243.26
98%
60°C to 63°C
20876-37-3
MFCD00100652
PBSZHNXXFIYDBU-UHFFFAOYSA-N
2-methyl-1-nitro-3-phenylmethoxybenzene
292648
Powder
1-Benzyloxy-2-methyl-3-nitrobenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures