missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyloxy-2-methyl-3-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ H5589914

 View more versions of this product

Catalog No. AAH5589914



Specifications

Specifications

1-Benzyloxy-2-methyl-3-nitrobenzene
60°C to 63°C
20876-37-3
MFCD00100652
PBSZHNXXFIYDBU-UHFFFAOYSA-N
2-methyl-1-nitro-3-phenylmethoxybenzene
292648
Powder
Brown to Yellow
25g
C14H13NO3
1-benzyloxy-2-methyl-3-nitrobenzene
CC1=C(C=CC=C1OCC2=CC=CC=C2)[N+](=O)[O-]
243.262
243.26
98%

Chemical Identifiers

20876-37-3
243.262
PBSZHNXXFIYDBU-UHFFFAOYSA-N
292648
CC1=C(C=CC=C1OCC2=CC=CC=C2)[N+](=O)[O-]
C14H13NO3
MFCD00100652
1-benzyloxy-2-methyl-3-nitrobenzene
2-methyl-1-nitro-3-phenylmethoxybenzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents