Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzylindole-5-boronic acid pinacol ester, 95%

$81.51 - $81.51

Chemical Identifiers

Chemical Name or Material 1-Benzylindole-5-boronic acid pinacol ester
Molecular Formula C21H24BNO2
Formula Weight 333.24
MDL Number MFCD11894320
Synonym 1-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5194503

  SDS
alfa aesar™
H5194503
1g Each for $81.51

AAH5194506

  SDS
alfa aesar™
H5194506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 333.24
Quantity 1g
Assay 95%
MDL Number MFCD11894320
Synonym 1-Benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
Chemical Name or Material 1-Benzylindole-5-boronic acid pinacol ester
Molecular Formula C21H24BNO2