missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzylimidazole, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152250050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Benzylimidazole | |
98.5 | |
310.0°C | |
99% | |
C10H10N2 | |
5g | |
1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole | |
KKKDZZRICRFGSD-UHFFFAOYSA-N | |
1-benzylimidazole | |
77918 | |
99% |
4238-71-5 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00005296 | |
23, 48 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol and acetone | |
C1=CC=C(C=C1)CN2C=CN=C2 | |
158.2 | |
158.2 |
Chemical Identifiers
4238-71-5 | |
158.2 | |
KKKDZZRICRFGSD-UHFFFAOYSA-N | |
77918 | |
C1=CC=C(C=C1)CN2C=CN=C2 |
C10H10N2 | |
MFCD00005296 | |
1-benzyl-1h-imidazole, n-benzylimidazole, imidazole, 1-benzyl, benzylimidazole, 1h-imidazole, 1-phenylmethyl, 1-benzyl imidazole, ccris 5821, chembl14192, 1bn, 1-benzylimdazole | |
1-benzylimidazole |
Safety and Handling
EINECSNumber : 224-200-4
RUO â Research Use Only