missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzylbenzimidazole, 97%

$41.01 - $181.97

Chemical Identifiers

CAS 141437-85-6
Molecular Formula C14H21NO2S
Molecular Weight (g/mol) 267.387
MDL Number MFCD00458798
InChI Key PYBYEBRVJRDWPB-LBPRGKRZSA-N
Synonym s-2-benzyl-2-n-bocamino-ethyl thiol, s-tert-butyl 1-mercapto-3-phenylpropan-2-yl carbamate, tert-butyl n-2s-1-phenyl-3-sulfanylpropan-2-yl carbamate, s-2-benzyl-2-n-bocaminoethyl thiol, s-2-benzyl-2-n-boc-aminoethyl thiol, s-2-n-boc-2-amino-3-phenylpropane-1-thiol, s-1-mercaptomethyl-2-phenylethyl carbamic acid tert-butyl ester
PubChem CID 15003862
IUPAC Name tert-butyl N-[(2S)-1-phenyl-3-sulfanylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6008803 View Documents Alfa Aesar™
H6008803
1g Each for $41.01
 
AAH6008806 View Documents Alfa Aesar™
H6008806
5g Each for $181.97
 
Specifications

Chemical Identifiers

141437-85-6
267.387
PYBYEBRVJRDWPB-LBPRGKRZSA-N
15003862
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CS
C14H21NO2S
MFCD00458798
s-2-benzyl-2-n-bocamino-ethyl thiol, s-tert-butyl 1-mercapto-3-phenylpropan-2-yl carbamate, tert-butyl n-2s-1-phenyl-3-sulfanylpropan-2-yl carbamate, s-2-benzyl-2-n-bocaminoethyl thiol, s-2-benzyl-2-n-boc-aminoethyl thiol, s-2-n-boc-2-amino-3-phenylpropane-1-thiol, s-1-mercaptomethyl-2-phenylethyl carbamic acid tert-butyl ester
tert-butyl N-[(2S)-1-phenyl-3-sulfanylpropan-2-yl]carbamate

Specifications

108°C to 110°C
141437-85-6
MFCD00458798
PYBYEBRVJRDWPB-LBPRGKRZSA-N
tert-butyl N-[(2S)-1-phenyl-3-sulfanylpropan-2-yl]carbamate
15003862
97%
1g
C14H21NO2S
s-2-benzyl-2-n-bocamino-ethyl thiol, s-tert-butyl 1-mercapto-3-phenylpropan-2-yl carbamate, tert-butyl n-2s-1-phenyl-3-sulfanylpropan-2-yl carbamate, s-2-benzyl-2-n-bocaminoethyl thiol, s-2-benzyl-2-n-boc-aminoethyl thiol, s-2-n-boc-2-amino-3-phenylpropane-1-thiol, s-1-mercaptomethyl-2-phenylethyl carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CS
267.387
208.26
1-Benzylbenzimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning