missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl L-Aspartate, TCI America™

$67.22 - $200.14

Chemical Identifiers

CAS 7362-93-8
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00063186
InChI Key NJSRYBIBUXBNSW-VIFPVBQESA-N
Synonym h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
PubChem CID 6994684
IUPAC Name (3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29941G View Documents TCI America
B29941G
1g Each for $67.22
 
B29945G View Documents TCI America
B29945G
5g Each for $200.14
 
Specifications

Chemical Identifiers

7362-93-8
223.228
NJSRYBIBUXBNSW-VIFPVBQESA-N
6994684
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
C11H13NO4
MFCD00063186
h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
(3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid

Specifications

White-Yellow
1g
C11H13NO4
h-asp-obzl, l-aspartic acid benzyl ester, l-aspartic acid 1-benzyl ester, s-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyl l-aspartate, 3s-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3s-3-amino-4-benzyloxy-4-oxobutanoic acid, h-asp-obzl.hcl, h-asp obzl, pubchem14915
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
223.228
223.23
≥98.0% (HPLC,T)
166°C
7362-93-8
MFCD00063186
NJSRYBIBUXBNSW-VIFPVBQESA-N
(3S)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
6994684
Crystal-Powder at 20°C
1-Benzyl L-Aspartate
Safety and Handling

Safety and Handling

TSCA : No