missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl D-Aspartate, TCI America™

$94.72 - $281.11

Chemical Identifiers

CAS 79337-40-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
InChI Key NJSRYBIBUXBNSW-SECBINFHSA-N
Synonym 1-benzyl d-aspartate, d-aspartic acid alpha-benzyl ester, 3r-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3r-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyld-aspartate, d-aspartic acid, a-benzyl ester, d-aspartic acid 1-benzyl ester, d-aspartic acid, 1-phenylmethyl ester
PubChem CID 6995008
IUPAC Name (3R)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29931G View Documents TCI America
B29931G
1g Each for $94.72
 
B29935G View Documents TCI America
B29935G
5g Each for $281.11
 
Specifications

Chemical Identifiers

79337-40-9
223.228
1-benzyl d-aspartate, d-aspartic acid alpha-benzyl ester, 3r-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3r-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyld-aspartate, d-aspartic acid, a-benzyl ester, d-aspartic acid 1-benzyl ester, d-aspartic acid, 1-phenylmethyl ester
(3R)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
C11H13NO4
NJSRYBIBUXBNSW-SECBINFHSA-N
6995008
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N

Specifications

White
1g
C11H13NO4
NJSRYBIBUXBNSW-SECBINFHSA-N
(3R)-3-amino-4-oxo-4-phenylmethoxybutanoic acid
6995008
Crystal-Powder at 20°C
1-Benzyl D-Aspartate
176°C
79337-40-9
1-benzyl d-aspartate, d-aspartic acid alpha-benzyl ester, 3r-3-amino-4-oxo-4-phenylmethoxybutanoic acid, 3r-3-amino-4-benzyloxy-4-oxobutanoic acid, 1-benzyld-aspartate, d-aspartic acid, a-benzyl ester, d-aspartic acid 1-benzyl ester, d-aspartic acid, 1-phenylmethyl ester
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)N
223.228
223.23
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No