Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-7-azaindole-3-boronic acid pinacol ester, 95%

$87.74 - $87.74

Chemical Identifiers

Chemical Name or Material 1-Benzyl-7-azaindole-3-boronic acid pinacol ester
Molecular Formula C20H23BN2O2
Formula Weight 334.22
MDL Number MFCD16294550
Synonym 1-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5194903

 
alfa aesar™
H5194903
1g Each for $87.74

AAH5194906

 
alfa aesar™
H5194906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 334.22
Chemical Name or Material 1-Benzyl-7-azaindole-3-boronic acid pinacol ester
Assay 95%
Molecular Formula C20H23BN2O2
MDL Number MFCD16294550
Synonym 1-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Quantity 1g