missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-4-piperidone, TCI America™

$39.72 - $397.23

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B102725ML View Documents TCI America
B102725ML
25mL Each for $39.72
 
B1027500ML View Documents TCI America
B1027500ML
500mL Each for $397.23
 
Specifications

Chemical Identifiers

3612-20-2
189.258
SJZKULRDWHPHGG-UHFFFAOYSA-N
19220
C1CN(CCC1=O)CC2=CC=CC=C2
C12H15NO
MFCD00006192
1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
1-benzylpiperidin-4-one

Specifications

158°C
25mL
C12H15NO
1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
C1CN(CCC1=O)CC2=CC=CC=C2
189.258
189.26
≥98.0% (GC,T)
Yellow
3612-20-2
MFCD00006192
SJZKULRDWHPHGG-UHFFFAOYSA-N
1-benzylpiperidin-4-one
19220
Clear Liquid at 20°C
1-Benzyl-4-piperidone
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3734

TSCA : No

Recommended Storage : Refrigerator