Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-piperidone, 98+%

$44.45 - $530.38

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-piperidone
CAS 3612-20-2
Molecular Formula C12H15NO
Formula Weight 189.26
MDL Number MFCD00006192
Beilstein 128556
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1239036

  SDS
alfa aesar™
A1239036
500g Each for $530.38

AAA1239014

  SDS
alfa aesar™
A1239014
25g Each for $44.45
Description & Specifications

Specifications

Formula Weight 189.26
Chemical Name or Material 1-Benzyl-4-piperidone
Assay ≥98%
Molecular Formula C12H15NO
MDL Number MFCD00006192
CAS 3612-20-2
Beilstein 128556
Boiling Point 248°C
Quantity 500g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2°C to 8°C

Applications
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

Solubility
Soluble in water (12 g/100ml at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

2°C to 8°C